Anti-CA1 Goat antibody

Supplier: OriGene
ORIGTA305634EA 454 EUR
ORIGTA305634
Anti-CA1 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: CA1 (carbonic anhydrase I)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR