Anti-WNT9B Goat Antibody

Supplier: OriGene
ORIGTA305646EA 454 EUR
ORIGTA305646
Anti-WNT9B Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: WNT9B (wingless-type MMTV integration site family, member 9B)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR