Anti-HMGA2 Goat Antibody

Supplier: OriGene
ORIGTA305648EA 547 EUR
ORIGTA305648
Anti-HMGA2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: HMGA2 (high mobility group AT-hook 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR