Anti-DLL1 Goat antibody

Supplier: OriGene
ORIGTA305657EA 547 EUR
ORIGTA305657
Anti-DLL1 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: DLL1 (delta-like 1 (Drosophila))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR