Anti-THY1 Goat Antibody

Supplier: OriGene
ORIGTA305663EA 547 EUR
ORIGTA305663
Anti-THY1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR