Anti-AGO1 Goat Antibody

Supplier: OriGene
ORIGTA305672EA 454 EUR
ORIGTA305672
Anti-AGO1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: AGO1 (argonaute RISC catalytic component 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR