Anti-MED15 Goat Antibody

Supplier: OriGene
ORIGTA305676EA 547 EUR
ORIGTA305676
Anti-MED15 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: MED15 (mediator complex subunit 15)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR