Anti-STK11IP Goat Antibody

Supplier: OriGene
ORIGTA305695EA 454 EUR
ORIGTA305695
Anti-STK11IP Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: STK11IP (serine/threonine kinase 11 interacting protein)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR