Anti-UNC13D Goat Antibody

Supplier: OriGene
ORIGTA305698EA 547 EUR
ORIGTA305698
Anti-UNC13D Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: UNC13D (unc-13 homolog D (C. elegans))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR