Anti-SPRY1 Goat Antibody

Supplier: OriGene
ORIGTA305701EA 456 EUR
ORIGTA305701
Anti-SPRY1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: SPRY1 (sprouty homolog 1, antagonist of FGF signaling (Drosophila))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR