Anti-PPP2R4 Goat Antibody

Supplier: OriGene
ORIGTA305706EA 500 EUR
ORIGTA305706
Anti-PPP2R4 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2?g/ml

Type: Primary
Antigen: PPP2R4 (protein phosphatase 2A activator, regulatory subunit 4)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR