Anti-CAMK2A Goat Antibody

Supplier: OriGene
ORIGTA305709EA 502 EUR
ORIGTA305709
Anti-CAMK2A Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 01-03?g/ml; Immunochemistry: 3-6 ?g/ml

Type: Primary
Antigen: CAMK2A (calcium/calmodulin-dependent protein kinase II alpha)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR