Anti-COG7 Goat Antibody

Supplier: OriGene
ORIGTA305712EA 502 EUR
ORIGTA305712
Anti-COG7 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: COG7 (component of oligomeric golgi complex 7)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR