Anti-MARCH10 Goat Antibody

Supplier: OriGene
ORIGTA305715EA 547 EUR
ORIGTA305715
Anti-MARCH10 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: MARCH10 (membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR