Anti-LPIN3 Goat Antibody

Supplier: OriGene
ORIGTA305728EA 547 EUR
ORIGTA305728
Anti-LPIN3 Goat Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 2-3?g/ml

Type: Primary
Antigen: LPIN3 (lipin 3)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR