Anti-ANLN Goat Antibody

Supplier: OriGene
ORIGTA305739EA 454 EUR
ORIGTA305739
Anti-ANLN Goat Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 3-10mg/ml

Type: Primary
Antigen: ANLN (anillin, actin binding protein)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR