Anti-SERPINA10 Goat Antibody

Supplier: OriGene
ORIGTA305744EA 547 EUR
ORIGTA305744
Anti-SERPINA10 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: SERPINA10 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR