Anti-STXBP6 Goat Antibody

Supplier: OriGene
ORIGTA305747EA 454 EUR
ORIGTA305747
Anti-STXBP6 Goat Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: STXBP6 (syntaxin binding protein 6 (amisyn))
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR