Anti-RAMP1 Goat Antibody

Supplier: OriGene
ORIGTA305755EA 454 EUR
ORIGTA305755
Anti-RAMP1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03?g/ml

Type: Primary
Antigen: RAMP1 (receptor (G protein-coupled) activity modifying protein 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR