Anti-PGLYRP1 Goat Antibody

Supplier: OriGene
ORIGTA305756EA 547 EUR
ORIGTA305756
Anti-PGLYRP1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml; Immunochemistry: 3-6?g/ml

Type: Primary
Antigen: PGLYRP1 (peptidoglycan recognition protein 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR