Anti-ABCD4 Goat Antibody

Supplier: OriGene
ORIGTA305757EA 547 EUR
ORIGTA305757
Anti-ABCD4 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: ABCD4 (ATP-binding cassette, sub-family D (ALD), member 4)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR