Anti-BAG5 Goat antibody

Supplier: OriGene
ORIGTA305758EA 550 EUR
ORIGTA305758
Anti-BAG5 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: BAG5 (BCL2-associated athanogene 5)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR