Anti-FMR1 Goat antibody

Supplier: OriGene
ORIGTA305783EA 547 EUR
ORIGTA305783
Anti-FMR1 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: FMR1 (fragile X mental retardation 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR