Anti-FKBP4 Goat antibody

Supplier: OriGene
ORIGTA305784EA 500 EUR
ORIGTA305784
Anti-FKBP4 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 001-003?g/ml

Type: Primary
Antigen: FKBP4 (FK506 binding protein 4, 59kDa)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR