Anti-ADRA2A Goat Antibody

Supplier: OriGene
ORIGTA305798EA 547 EUR
ORIGTA305798
Anti-ADRA2A Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1?g/ml Immunofluorescence: 3-5?g/ml

Type: Primary
Antigen: ADRA2A (adrenoceptor alpha 2A)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR