Anti-ERC1 Goat Antibody

Supplier: OriGene
ORIGTA305812EA 500 EUR
ORIGTA305812
Anti-ERC1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: ERC1 (ELKS/RAB6-interacting/CAST family member 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR