Anti-BAZ2B Goat Antibody

Supplier: OriGene
ORIGTA305814EA 550 EUR
ORIGTA305814
Anti-BAZ2B Goat Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: BAZ2B (bromodomain adjacent to zinc finger domain, 2B)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR