Anti-D130043K22Rik Goat Antibody

Supplier: OriGene
ORIGTA305831EA 500 EUR
ORIGTA305831
Anti-D130043K22Rik Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 001-003?g/ml

Type: Primary
Antigen: D130043K22Rik
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR