Anti-ACTL7B Goat Antibody

Supplier: OriGene
ORIGTA305837EA 547 EUR
ORIGTA305837
Anti-ACTL7B Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: ACTL7B (actin-like 7B)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR