Anti-APOC1 Goat Antibody

Supplier: OriGene
ORIGTA305838EA 500 EUR
ORIGTA305838
Anti-APOC1 Goat Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: APOC1 (apolipoprotein C-I)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR