Anti-DHX58 Goat Antibody

Supplier: OriGene
ORIGTA305842EA 454 EUR
ORIGTA305842
Anti-DHX58 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: DHX58 (DEXH (Asp-Glu-X-His) box polypeptide 58)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR