Anti-MC5R Goat Antibody

Supplier: OriGene
ORIGTA305847EA 550 EUR
ORIGTA305847
Anti-MC5R Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: MC5R (melanocortin 5 receptor)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR