Anti-ERAP2 Goat Antibody

Supplier: OriGene
ORIGTA305860EA 547 EUR
ORIGTA305860
Anti-ERAP2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: ERAP2 (endoplasmic reticulum aminopeptidase 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR