Anti-ABCC11 Goat Antibody

Supplier: OriGene
ORIGTA305865EA 454 EUR
ORIGTA305865
Anti-ABCC11 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: ABCC11 (ATP-binding cassette, sub-family C (CFTR/MRP), member 11)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR