Anti-ZRANB1 Goat Antibody

Supplier: OriGene
ORIGTA305868EA 502 EUR
ORIGTA305868
Anti-ZRANB1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ZRANB1 (zinc finger, RAN-binding domain containing 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR