Anti-ABCC1 Goat Antibody

Supplier: OriGene
ORIGTA305869EA 547 EUR
ORIGTA305869
Anti-ABCC1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot:005-02?g/ml

Type: Primary
Antigen: ABCC1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR