Anti-AVPR1B Goat Antibody

Supplier: OriGene
ORIGTA305874EA 498 EUR
ORIGTA305874
Anti-AVPR1B Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 005-02?g/ml

Type: Primary
Antigen: AVPR1B (arginine vasopressin receptor 1B)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR