Anti-CERS1 Goat Antibody

Supplier: OriGene
ORIGTA305879EA 500 EUR
ORIGTA305879
Anti-CERS1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: CERS1 (ceramide synthase 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR