Anti-TENM3 Goat Antibody

Supplier: OriGene
ORIGTA305884EA 454 EUR
ORIGTA305884
Anti-TENM3 Goat Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 15-3?g/ml Immunofluorescence: 10?g/ml

Type: Primary
Antigen: TENM3 (teneurin transmembrane protein 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR