Anti-CLU Goat Antibody

Supplier: OriGene
ORIGTA305888EA 547 EUR
ORIGTA305888
Anti-CLU Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: CLU (clusterin)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR