Anti-DPM1 Goat Antibody

Supplier: OriGene
ORIGTA305890EA 454 EUR
ORIGTA305890
Anti-DPM1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: DPM1 (dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR