Anti-ORAI1 Goat Antibody

Supplier: OriGene
ORIGTA305897EA 454 EUR
ORIGTA305897
Anti-ORAI1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: ORAI1 (ORAI calcium release-activated calcium modulator 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR