Anti-BTLA Goat Antibody

Supplier: OriGene
ORIGTA305899EA 454 EUR
ORIGTA305899
Anti-BTLA Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: BTLA (B and T lymphocyte associated)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR