Anti-TSPO Goat Antibody

Supplier: OriGene
ORIGTA305904EA 454 EUR
ORIGTA305904
Anti-TSPO Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 02-05?g/ml

Type: Primary
Antigen: TSPO (translocator protein (18kDa))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR