Anti-SH2D3A Goat Antibody

Supplier: OriGene
ORIGTA305906EA 456 EUR
ORIGTA305906
Anti-SH2D3A Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: SH2D3A (SH2 domain containing 3A)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR