Anti-HMGB3 Goat Antibody

Supplier: OriGene
ORIGTA305908EA 454 EUR
ORIGTA305908
Anti-HMGB3 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 001-003?g/ml

Type: Primary
Antigen: HMGB3 (high mobility group box 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR