Anti-CDK10 Goat Antibody

Supplier: OriGene
ORIGTA305909EA 456 EUR
ORIGTA305909
Anti-CDK10 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2?g/ml

Type: Primary
Antigen: CDK10 (cyclin-dependent kinase 10)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR