Anti-TEAD2 Goat Antibody

Supplier: OriGene
ORIGTA305910EA 547 EUR
ORIGTA305910
Anti-TEAD2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: TEAD2 (TEA domain family member 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR