Anti-F2R Goat Antibody

Supplier: OriGene
ORIGTA305911EA 547 EUR
ORIGTA305911
Anti-F2R Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2?g/ml

Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR