Anti-PPP4C Goat Antibody

Supplier: OriGene
ORIGTA305920EA 454 EUR
ORIGTA305920
Anti-PPP4C Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: PPP4C (protein phosphatase 4, catalytic subunit)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR